4-Nitrophenyl chloroformate 97%

Danger

AC17080-0050 AC17080-5000
200005-520EA 42.5 USD
200005-520 103526-288
4-Nitrophenyl chloroformate 97%
4-Nitrophenyl chloroformate
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 159…162 °C (19 mmHg)
Melting Pt: 73…81 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007321
CAS Number: 7693-46-1
EINECS: 231-706-9
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR