N,N,N',N'-Tetramethyl-p-phenylenediamine 98%

Warning

Synonyms: N,N,N,N-Tetramethyl-1,4-phenylenediamineTMPDN,N,N',N'-Tetramethyl-1,4-phenylenediamine

AC17079-1000 AC17079-0050 AC17079-0250
103531-656EA 889 USD
103531-656 200013-500 200038-038
N,N,N',N'-Tetramethyl-p-phenylenediamine 98%
N,N,N',N'-Tetramethyl-p-phenylenediamine
Formula: C₁₀H₁₆N₂
MW: 164.25 g/mol
Boiling Pt: 260 °C (760 mmHg)
Melting Pt: 48…52 °C
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008309
CAS Number: 100-22-1
EINECS: 202-831-6
UN: 2811
ADR: 6.1,III
Merck Index: 12,09367

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR