tert-Butyl perbenzoate 98%

Danger

Synonyms: Luperox(R) P, tert-Butyl peroxybenzoatetert-Butyl peroxybenzoate

AC17014-0010
103531-640EA 133 USD
103531-640
tert-Butyl perbenzoate 98%
tert-Butyl perbenzoate
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 75…76 °C (.2 mmHg)
Melting Pt: 8 °C
Density: 1.04 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00008802
CAS Number: 614-45-9
EINECS: 210-382-2
UN: 3103
ADR: 5.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR