Pentafluoropyridine 99+%

Danger

Synonyms: 2,3,4,5,6-Perfluoropyridine2,3,4,5,6-Pentafluoropyridine

AC16999-0250 AC16999-0050
200026-362EA 41.2 USD
200026-362 200050-406
Pentafluoropyridine 99+%
Pentafluoropyridine
Formula: C₅F₅N
MW: 169.05 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: –41 °C
Density: 1.54 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00006225
CAS Number: 700-16-3
EINECS: 211-839-9
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR