4-Methylquinoline 99%

Warning

Synonyms: Lepidine

AC16977-1000 AC16977-0050 AC16977-0250
200025-680EA 36.4 USD
200025-680 200043-768 200037-788
4-Methylquinoline 99%
4-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.08 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006784
CAS Number: 491-35-0
EINECS: 207-734-2
Merck Index: 13,05460

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR