(±)-3-Methylcyclopentanone 99%

Warning

Synonyms: DL-3-Methylcyclopentanone

AC16937-0050 AC16937-0250
103531-634EA 290 USD
103531-634 200025-166
(±)-3-Methylcyclopentanone 99%
(±)-3-Methylcyclopentanone
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 145 °C (760 mmHg)
Density: 0.91 g/cm³
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00001416
CAS Number: 1757-42-2
EINECS: 217-148-9
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR