Heptafluorobutyric anhydride (HFAA) 98+%

Danger

Synonyms: Perfluorobutyric anhydride2,2,3,3,4,4,4-Heptafluorobutanoic anhydrideHFBAHFAA

AC16926-1000 AC16926-0050 AC16926-0250
200001-650EA 156 USD
200001-650 200012-640 103526-252
Heptafluorobutyric anhydride (HFAA) 98+%
Heptafluorobutyric anhydride
Formula: C₈F₁₄O₃
MW: 410.06 g/mol
Boiling Pt: 108…111 °C (760 mmHg)
Melting Pt: –43 °C
Density: 1.65 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000432
CAS Number: 336-59-4
EINECS: 206-410-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR