Phenanthridine 98%

Warning

AC16925-0050 AC16925-0010
200044-416EA 118 USD
200044-416 200032-502
Phenanthridine 98%
Phenanthridine
Formula: C₁₃H₉N
Boiling Pt: 349 °C (769 mmHg)
Melting Pt: 104…108 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004989
CAS Number: 229-87-8
EINECS: 205-934-4
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR