L(+)-Diethyl tartrate 99+%

Synonyms: (R,R)-(+)-Diethyl tartrate(+)-Diethyl L-tartrateDiethyl L-tartrate(2R,3R)-Diethyl 2,3-dihydroxysuccinateL(+)-Tartaric acid diethyl ester(R,R)-(+)-Tartaric acid diethyl ester(2R,3R)-(+)-Diethyl tartrate

AC16896-1000 AC16896-5000
200004-498EA 66.6 USD
200004-498 200008-130
L(+)-Diethyl tartrate 99+%
L(+)-Diethyl tartrate
Formula: C₈H₁₄O₆
MW: 206.2 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.2 g/cm³
Flash Pt: 93 °C
MDL Number: MFCD00009143
CAS Number: 87-91-2
EINECS: 201-783-3
Merck Index: 13,03886

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR