Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%

Warning

Synonyms: 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diolBis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol

AC16887-0025 AC16887-1000 AC16887-5000
103526-244EA 412 USD
103526-244 103526-242 200001-642
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)
Formula: C₈H₁₉NO₅
MW: 209.24 g/mol
Melting Pt: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002853
CAS Number: 6976-37-0
EINECS: 230-237-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR