Cyclobutanol 99+%

Danger

AC16875-0050 AC16875-0010 AC16875-2500
103531-624EA 315 USD
103531-624 103531-626 200011-810
Cyclobutanol 99+%
Cyclobutanol
Formula: C₄H₈O
Boiling Pt: 123…124 °C (733 mmHg)
Density: 0.921 g/cm³
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00001318
CAS Number: 2919-23-5
EINECS: 220-858-1
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR