Ammonium formate ≥97%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: Formic acid ammonium salt

AC16861-0051 AC16861-5000 AC16861-0010
103464-006EA 377.55 USD InStock
103464-006 200011-216 200005-742
Ammonium formate ≥97%
Ammonium formate
Formula: HCO₂NH₄
MW: 63.06 g/mol
Melting Pt: 119 - 121 °C
Density: 1.28 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR