Sodium ethoxide ≥96%, pure

Supplier: ACROS ORGANICS MS

Danger

Synonyms: Sodium ethylateSodium ethanolate

AC16859-2500 AC16859-0025 AC16859-5000
200001-638EA 68.03 USD InStock
200001-638 103526-230 200001-640
Sodium ethoxide ≥96%, pure
Sodium ethoxide
Formula: C₂H₅NaO
MW: 68.05 g/mol
Melting Pt: 260 °C
Density: 0.868 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3095
ADR: 4.2,II

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Alkalinity ≥98 % (Total)
Titration ≥95 %
NaOH + Na2CO3 (as NaOH) ≤2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR