Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy

Danger

Synonyms: Ethylene glycol monomethyl ether

AC16858-0010 AC16858-0025
200041-134EA 450 USD
200041-134 200009-462
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124.6 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.96 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR