Sodium cyanotrihydroborate 95%

Danger

Synonyms: Sodium cyanoborohydride

AC16855-0100 AC16855-2500 AC16855-0500
200001-634EA 60.2 USD
200001-634 103526-228 200001-636
Sodium cyanotrihydroborate 95%
Sodium cyanotrihydroborate
Formula: CH₃BNNa
MW: 62.84 g/mol
Melting Pt: >242 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003516
CAS Number: 25895-60-7
EINECS: 247-317-2
UN: 3134
ADR: 4.3,II
Merck Index: 12,08751

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR