Methyl-3-nitrobenzoate 98%

Synonyms: Methyl 3-Nitrobenzoate3-Nitrobenzoic acid methyl ester

AC16844-1000 AC16844-5000 AC16844-0250
103510-926EA 195 USD
103510-926 200025-840 103510-924
Methyl-3-nitrobenzoate 98%
Methyl 3-nitrobenzoate
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 279 °C (760 mmHg)
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00007250
CAS Number: 618-95-1
EINECS: 210-574-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR