Ethyl-4-nitrobenzoate 99%

Synonyms: 4-Nitrobenzoic acid ethyl esterEthyl 4-nitrobenzoate

AC16842-1000 AC16842-5000
200037-372EA 408 USD
200037-372 200025-314
Ethyl-4-nitrobenzoate 99%
Ethyl 4-nitrobenzoate
Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00007351
CAS Number: 99-77-4
EINECS: 202-786-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR