2-Fluoro-5-nitroaniline 98%

Warning

AC16839-0250 AC16839-1000 AC16839-0050
200016-556EA 172 USD
200016-556 103531-622 200034-800
2-Fluoro-5-nitroaniline 98%
2-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 99…103 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR