(±)-2-Methyltetrahydrofuran 99+% stabilized, pure

Danger

AC16836-0100 AC16836-1000 AC16836-0010 AC16836-5000 AC16836-0025
103526-552EA 969.23 USD
103526-552 200005-124 103526-222 103526-224 200001-628
(±)-2-Methyltetrahydrofuran 99+% stabilized, pure
(±)-2-Methyltetrahydrofuran
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 78…80 °C (760 mmHg)
Melting Pt: –136 °C
Density: 0.86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR