2,2,6,6-Tetramethyl-3,5-heptanedione 98%

Warning

Synonyms: DipivaloylmethaneTMHD2,2,6,6-Tetramethylheptane-3,5-dione

AC16832-0050 AC16832-1000 AC16832-0250
200008-868EA 157 USD
200008-868 103510-922 200022-028
2,2,6,6-Tetramethyl-3,5-heptanedione 98%
2,2,6,6-Tetramethyl-3,5-heptanedione
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 72…73 °C (6 mmHg)
Density: 0.88 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008848
CAS Number: 1118-71-4
EINECS: 214-268-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR