2-Chlorothiophene

Synonyms: 2-Thienyl chloride

AC16826-0250 AC16826-1000
200040-988EA 72.4 USD
200040-988 200022-600
2-Chlorothiophene
2-Chlorothiophene
Formula: C₄H₃ClS
MW: 118.59 g/mol
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –1 °C
Density: 1.285 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005421
CAS Number: 96-43-5
EINECS: 202-505-3
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR