Valeronitrile 98%

Warning

Synonyms: Pentanenitrile

AC16794-5000 AC16794-1000
200008-054EA 65.8 USD
200008-054 103531-610
Valeronitrile 98%
Valeronitrile
Formula: C₅H₉N
Boiling Pt: 139…141 °C (760 mmHg)
Melting Pt: –96 °C
Density: 0.79 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00001974
CAS Number: 110-59-8
EINECS: 203-781-8
UN: 1992
ADR: 3,III
Merck Index: 12,10043

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR