3-Fluorophenylhydrazinium chloride 97%

Danger

Synonyms: (3-Fluorphenyl)hydrazin HCl3-Fluorophenylhydrazine hydrochloride1-(3-Fluorophenyl)hydrazine hydrochloride(3-Fluorophenyl)hydrazine hydrochloride

AC16737-0100 AC16737-0050
200029-300EA 115 USD
200029-300 200047-476
3-Fluorophenylhydrazinium chloride 97%
3-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
Melting Pt: 268 °C (dec.)
MDL Number: MFCD00012934
CAS Number: 2924-16-5
EINECS: 220-886-4
UN: 2811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR