2-Adamantanol 98%

Synonyms: 2-Tricyclo[3.3.1.1(3,7)]decanolTricyclo[3.3.1.1(3,7)]decan-2-ol2-Hydroxytricyclo[3.3.1.1(3,7)]decane

AC16731-0050
200022-372EA 68.3 USD
200022-372
2-Adamantanol 98%
2-Adamantanol
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Melting Pt: 258…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00074744
CAS Number: 700-57-2
EINECS: 211-846-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR