Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-α,α,α-trifluorotoluene ≥99%

Supplier: ACROS ORGANICS MS

Warning
Synonyms: 4-Bromobenzotrifluoride1-Bromo-4-(trifluoromethyl)benzene

AC16667-0250 AC16667-0050 AC16667-1000
200017-404EA 189.67 USD InStock
200017-404 200010-258 103531-590
4-Bromo-α,α,α-trifluorotoluene ≥99%
4-Bromo-α,α,α-trifluorotoluene


Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 154…155 °C (760 mmHg)
Density: 1.6347 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00000398
CAS Number: 402-43-7
EINECS: 206-943-6
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4715 to 1.4745 (20°C, 589 nm)
GC ≥98.5 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty