Pyridine-[D₅] 99 atom % D for NMR spectroscopy

Danger

Synonyms: Pyridine-[²H₅]

AC16657-0250 AC16657-0050 AC16657-0100
200014-292EA 262 USD
200014-292 200026-638 200014-290
Pyridine-[D₅] 99 atom % D for NMR spectroscopy
Pyridine-[D5]
Formula: C₅D₅N
MW: 84.06 g/mol
Boiling Pt: 114.4 °C (1013 hPa)
Melting Pt: –41 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00044639
CAS Number: 7291-22-7
EINECS: 230-720-2
UN: 1282
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR