Dimethyl sulfoxide-[D₆] 99.9 atom % D for NMR spectroscopy

Synonyms: Dimethyl sulfoxide-[²H₆]Hexadeuterodimethyl sulfoxideDMSO-D6DMSO-d6Dimethyl sulphoxide-d6Dimethyl sulfoxide-d6Dimethyl sulphoxide-[²H₆]Hexadeuterodimethyl sulphoxide

AC16629-1000 AC16629-0500 AC16629-0100
200001-550EA 186 USD
200001-550 200001-552 200001-548
Dimethyl sulfoxide-[D₆] 99.9 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: 189 °C (5 mmHg)
Melting Pt: 20.2 °C
Density: 1.19 g/cm³
Flash Pt: 88 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR