2,4-Dimethylbenzaldehyde 99%

AC16579-1000 AC16579-0250 AC16579-0050
200022-198EA 46 USD
200022-198 103510-854 200022-200
2,4-Dimethylbenzaldehyde 99%
2,4-Dimethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 102.4…103 °C (14 mmHg)
Melting Pt: –9 °C
Density: 0.96 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003340
CAS Number: 15764-16-6
EINECS: 239-856-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR