5α-Cholestane 98+%

AC16560-0010 AC16560-0050 AC16560-2500
200018-500EA 63.39 USD
200018-500 103510-850 200024-744
5α-Cholestane 98+%
5α-Cholestane
Formula: C₂₇H₄₈
MW: 372.68 g/mol
Boiling Pt: 250 °C (1 mmHg)
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00066412
CAS Number: 481-21-0
EINECS: 207-562-8
Merck Index: 12,02254

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR