Dimethyl disulfide 99%
|
Danger
|
Synonyms:
Dimethyl disulfide
, Dimethyl disulphide
, Methyl disulphide
, Methyl disulfide
AC16559-0010
AC16559-2500
AC16559-0025
AC16559-0500
200006-988EA
36.5
USD
200006-988
103526-148
200001-502
103526-150
Dimethyl disulfide 99%
Dimethyl disulphide
Formula:
C₂H₆S₂ MW: 94.2 g/mol Boiling Pt: 108…110 °C (760 mmHg) Melting Pt: –85 °C Density: 1.0625 g/cm³ Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008561 CAS Number: 624-92-0 EINECS: 210-871-0 UN: 2381 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...