Phenyl salicylate 99%

Warning

Synonyms: Phenyl SalicylatePhenyl 2-hydroxybenzoate

AC16508-0010 AC16508-2500 AC16508-0025
103466-298EA 290 USD
103466-298 200013-992 200013-994
Phenyl salicylate 99%
Phenyl salicylate
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 172…173 °C (12 mmHg)
Melting Pt: 40…44 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002213
CAS Number: 118-55-8
EINECS: 204-259-2
ADR: 9,III
Merck Index: 12,07464

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR