Methyl myristate 99%

Synonyms: Methyl tetradecanoateTetradecylic acid methyl esterMyristic acid methyl esterTetradecanoic acid methyl ester

AC16501-0025 AC16501-5000 AC16501-1000
200043-918EA 188 USD
200043-918 200019-736 200043-920
Methyl myristate 99%
Methyl myristate
Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 18.4 °C
Density: 0.863 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008983
CAS Number: 124-10-7
EINECS: 204-680-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR