Ethyl hexanoate 99+%

Warning

Synonyms: Ethyl caproateCaproic acid ethyl esterHexanoic acid ethyl ester

AC16499-0050 AC16499-5000 AC16499-1000
200031-274EA 22.4 USD
200031-274 103531-568 200019-108
Ethyl hexanoate 99+%
Ethyl hexanoate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 168 °C (760 mmHg)
Melting Pt: 67 °C
Density: 0.869 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00009511
CAS Number: 123-66-0
EINECS: 204-640-3
UN: 3272
ADR: 3,III
Merck Index: 13,03812

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR