2-Methoxynaphthalene 98%

Synonyms: Methyl 2-naphthyl ether

AC16475-5000 AC16475-0050
200016-650EA 186 USD
200016-650 200016-648
2-Methoxynaphthalene 98%
2-Methoxynaphthalene
Formula: C₁₁H₁₀O
MW: 158.2 g/mol
Boiling Pt: 274 °C (1013 hPa)
Melting Pt: 73…75 °C
MDL Number: MFCD00004061
CAS Number: 93-04-9
EINECS: 202-213-6
Merck Index: 12,06076

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR