4-Methylanisole 99%

Warning

Synonyms: 1-Methoxy-4-methylbenzenep-Methylanisole4-Cresol methyl ether4-Methoxytoluene

AC16466-0025 AC16466-2500 AC16466-0050 AC16466-0010
200017-630EA 48.8 USD
200017-630 103531-558 200023-918 200047-594
4-Methylanisole 99%
4-Methylanisole
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 174 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.96 g/cm³
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00008413
CAS Number: 104-93-8
EINECS: 203-253-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR