4,4'-Bis(dimethylamino)benzophenone 98%

Danger

Synonyms: 4,4-Bis(dimethylamino)benzophenone4,4′-Bis(dimethylamino)benzophenoneBis[4-(dimethylamino)phenyl]methanone4,4'-Di(dimethylamino)benzophenoneMichler's ketone

AC16449-0050 AC16449-5000 AC16449-1000
200035-598EA 228 USD
200035-598 200023-510 200023-512
4,4'-Bis(dimethylamino)benzophenone 98%
4,4'-Bis(dimethylamino)benzophenone
Formula: C₁₇H₂₀N₂O
MW: 268.36 g/mol
Boiling Pt: 428 °C (1013 hPa)
Melting Pt: 171…176 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00008312
CAS Number: 90-94-8
EINECS: 202-027-5
ADR: 6.1,III
Merck Index: 12,06265

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR