2-Indanone 97%

Synonyms: beta-Hydrindone2-Oxoindane1H-Inden-2(3H)-one

AC16411-0500 AC16411-0100
200009-436EA 288 USD
200009-436 200009-434
2-Indanone 97%
2-Indanone
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 220…225 °C (1013 hPa)
Melting Pt: 50…57 °C
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003792
CAS Number: 615-13-4
EINECS: 210-410-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR