1-Ethyl-2-pyrrolidone 98%

Danger

Synonyms: 1-Ethyl-2-pyrrolidinoneN-Ethyl-2-pyrrolidone1-Ethylpyrrolidin-2-one

AC16393-0250 AC16393-1000
200033-928EA 62.8 USD
200033-928 200033-930
1-Ethyl-2-pyrrolidone 98%
1-Ethyl-2-pyrrolidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 212.5 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.998 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003199
CAS Number: 2687-91-4
EINECS: 220-250-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR