3-Cyanobenzaldehyde 99%

Warning

Synonyms: 3-Formylbenzonitrile

AC16390-0250 AC16390-0050 AC16390-0010
200035-342EA 284 USD
200035-342 200035-340 103531-544
3-Cyanobenzaldehyde 99%
3-Cyanobenzaldehyde
Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00003344
CAS Number: 24964-64-5
EINECS: 246-549-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR