6-Hydroxy-m-toluic acid 98%

Warning

Synonyms: 5-Methylsalicylic acid2,5-Cresotic acid

AC16386-0100 AC16386-0500
200017-860EA 74.4 USD
200017-860 103465-628
6-Hydroxy-m-toluic acid 98%
6-Hydroxy-m-toluic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 151…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00002461
CAS Number: 89-56-5
EINECS: 201-918-6
Merck Index: 12,02650

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR