Quinine sulfate dihydrate 99+%

Warning

Synonyms: bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulphuric acid dihydrate(-)-Quinine sulfate dihydrate(-)-Quinine sulphate dihydratebis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate

AC16371-0100 AC16371-0500
200020-580EA 284 USD
200020-580 200007-560
Quinine sulfate dihydrate 99+%
(-)-Quinine sulphate dihydrate
Formula: C₄₀H₅₀N₄O₈S·2H₂O
MW: 782.96 g/mol
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00150790
CAS Number: 6119-70-6
EINECS: 212-359-2
Merck Index: 12,03822

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR