δ-Valerolactone 99%

Danger

Synonyms: δ-ValerolactoneTetrahydro-2H-pyran-2-one5-Hydroxypentanoic acid lactonePentan-5-olide

AC16271-2500 AC16271-0010 AC16271-0500
200024-926EA 240 USD
200024-926 103510-794 200006-216
δ-Valerolactone 99%
δ-Valerolactone
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 230 °C (760 mmHg)
Melting Pt: –13 °C
Density: 1.1 g/cm³
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006645
CAS Number: 542-28-9
EINECS: 208-807-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR