2-(Bromomethyl)naphthalene 96%

Danger

AC16256-0010 AC16256-1000 AC16256-0050 AC16256-0250
200001-482EA 21 USD
200001-482 103526-094 103526-092 103526-096
2-(Bromomethyl)naphthalene 96%
2-(Bromomethyl)naphthalene
Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 213 °C (100 mmHg)
Melting Pt: 52…57 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004123
CAS Number: 939-26-4
EINECS: 213-359-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR