2,2'-Dipyridyl disulfide 98%

Warning

Synonyms: 1,2-Di(pyridin-2-yl)disulfane1,2-Di(pyridin-2-yl)disulphane2,2'-Dithiodipyridine

AC16223-0250 AC16223-1000 AC16223-0050
200004-850EA 90.4 USD
200004-850 200022-052 103526-084
2,2'-Dipyridyl disulfide 98%
2,2'-Dipyridyl disulphide
Formula: C₁₀H₈N₂S₂
MW: 220.32 g/mol
Melting Pt: 56…60 °C
Flash Pt: 210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006287
CAS Number: 2127-03-9
EINECS: 218-343-1
UN: 3335
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR