Difluoroacetic acid 98%

Danger

AC16212-0500 AC16212-0100 AC16212-0025
200018-808EA 131.54 USD
200018-808 200025-032 103526-080
Difluoroacetic acid 98%
Difluoroacetic acid
Formula: C₂H₂F₂O₂
MW: 96.03 g/mol
Boiling Pt: 132…134 °C (760 mmHg)
Melting Pt: –1 °C
Density: 1.52 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004220
CAS Number: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR