Phenyl benzoate 99%

Warning

Synonyms: Benzoic acid phenyl ester

AC16206-5000 AC16206-0050
200032-516EA 24.4 USD
200032-516 200013-984
Phenyl benzoate 99%
Phenyl benzoate
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00003072
CAS Number: 93-99-2
EINECS: 202-293-2
Merck Index: 12,07429

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR