2,4,6-Collidine 99%

Danger

Synonyms: sym-Collidine2,4,6-Trimethylpyridine

AC16195-1000 AC16195-0010 AC16195-5000 AC16195-0025
103526-068EA 825 USD
103526-068 103526-066 200001-478 200016-064
2,4,6-Collidine 99%
2,4,6-Collidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 171…172 °C (760 mmHg)
Melting Pt: –43 °C
Density: 0.91 g/cm³
Flash Pt: 57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006338
CAS Number: 108-75-8
EINECS: 203-613-3
UN: 1992
ADR: 3,III
Merck Index: 12,09848

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR