2-Chloro-6-fluorobenzaldehyde 95%

Warning

AC16151-0250 AC16151-5000 AC16151-1000
200028-632EA 32.8 USD
200028-632 200028-634 103510-784
2-Chloro-6-fluorobenzaldehyde 95%
2-Chloro-6-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 92 °C (10 mmHg)
Melting Pt: 34…39 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00003306
CAS Number: 387-45-1
EINECS: 206-860-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR