3,5-Di-tert-butyl-4-hydroxybenzaldehyde 97%

Warning

AC16121-0250
200016-906EA 68.3 USD
200016-906
3,5-Di-tert-butyl-4-hydroxybenzaldehyde 97%
3,5-Di-tert-butyl-4-hydroxybenzaldehyde
Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 186…190 °C
MDL Number: MFCD00008826
CAS Number: 1620-98-0
EINECS: 216-592-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR