Veratrole 99+%

Warning

Synonyms: 1,2-DimethoxybenzenePyrocatechol dimethyl ether

AC16111-0010 AC16111-5000 AC16111-1000
200015-146EA 67 USD
200015-146 200015-144 200027-160
Veratrole 99+%
Veratrole
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.08 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00008357
CAS Number: 91-16-7
EINECS: 202-045-3
Merck Index: 12,10089

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR